Assay Details
| Gene Symbol: hui | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137147.5 | NP_610991.1 | 3 - 4 | 389 | 78 | |
| NM_166071.4 | NP_725421.1 | 2 - 3 | 476 | 78 | ||
| GenBank mRNA | AY118599.1 | - | 2 - 3 | 446 | 78 | |
More Information
Set Membership: |
|

hui
Genomic Map