Assay Details
| Gene Symbol: Hr51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137188.3 | NP_611032.2 | 7 - 8 | 1510 | 86 | |
| GenBank mRNA | BT122090.2 | - | 7 - 8 | 1234 | 86 | |
More Information
Set Membership: |
|

Hr51
Genomic Map