Assay Details
| Gene Symbol: Strn-Mlck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202026.2 | NP_001188955.2 | 8 - 9 | 17132 | 65 | |
| NM_001274089.1 | NP_001261018.1 | 8 - 9 | 17132 | 65 | ||
| NM_001274090.1 | NP_001261019.1 | 8 - 9 | 17500 | 65 | ||
| NM_166129.3 | NP_725510.2 | 8 - 9 | 17500 | 65 | ||
More Information
Set Membership: |
|

Strn-Mlck
Genomic Map