Assay Details
| Gene Symbol: Strn-Mlck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166130.2 | NP_725511.2 | 7 - 8 | 6205 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strn-Mlck| Gene Symbol: Strn-Mlck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166130.2 | NP_725511.2 | 7 - 8 | 6205 | 110 | |
Set Membership: |
|