Assay Details
| Gene Symbol: ATPCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144202.3 | NP_001137674.2 | 4 - 5 | 1181 | 81 | |
| NM_001299541.1 | NP_001286470.1 | 4 - 5 | 1271 | 81 | ||
| NM_079031.3 | NP_523755.2 | 4 - 5 | 1181 | 81 | ||
| NM_166131.3 | NP_725514.2 | 4 - 5 | 1271 | 81 | ||
| GenBank mRNA | AF132166.2 | - | 4 - 5 | 1130 | 81 | |
More Information
Set Membership: |
|

ATPCL
Genomic Map