Assay Details
| Gene Symbol: park | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168884.2 | NP_730600.1 | 2 - 3 | 350 | 113 | |
| NM_168885.3 | NP_730601.1 | 1 - 2 | 329 | 113 | ||
| GenBank mRNA | AY093423.3 | - | 1 - 2 | 271 | 113 | |
| BT044504.1 | - | 1 - 2 | 271 | 113 | ||
More Information
Set Membership: |
|

park
Genomic Map