Assay Details
| Gene Symbol: TfAP-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275250.1 | NP_001262179.1 | 1 - 2 | 262 | 68 | |
| NM_168927.2 | NP_730664.1 | 1 - 2 | 262 | 68 | ||
| GenBank mRNA | AJ006868.1 | - | 1 - 2 | 262 | 68 | |
More Information
Set Membership: |
|

TfAP-2
Genomic Map