Assay Details
| Gene Symbol: jim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014601.1 | NP_001014601.1 | 1 - 2 | 816 | 74 | |
| NM_001259959.2 | NP_001246888.1 | 2 - 3 | 1687 | 74 | ||
| NM_001275292.1 | NP_001262221.1 | 3 - 4 | 2781 | 74 | ||
| NM_001275293.1 | NP_001262222.1 | 2 - 3 | 1687 | 74 | ||
| NM_001275294.1 | NP_001262223.1 | 2 - 3 | 2344 | 74 | ||
| NM_143699.4 | NP_651956.1 | 3 - 4 | 2781 | 74 | ||
| NM_168964.5 | NP_730746.1 | 2 - 3 | 1687 | 74 | ||
| GenBank mRNA | AJ133756.1 | - | 2 - 3 | 2345 | 74 | |
| BT030442.1 | - | 1 - 2 | 506 | 74 | ||
More Information
Set Membership: |
|

jim
Genomic Map