Assay Details
| Gene Symbol: jim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014601.1 | NP_001014601.1 | 4 - 5 | 2248 | 75 | |
| NM_001275292.1 | NP_001262221.1 | 6 - 7 | 4213 | 75 | ||
| NM_001275294.1 | NP_001262223.1 | 5 - 6 | 3692 | 75 | ||
| NM_143699.4 | NP_651956.1 | 6 - 7 | 4213 | 75 | ||
| NM_168964.5 | NP_730746.1 | 5 - 6 | 3119 | 75 | ||
| GenBank mRNA | AJ133756.1 | - | 5 - 6 | 3777 | 75 | |
| BT030442.1 | - | 4 - 5 | 1938 | 75 | ||
More Information
Set Membership: |
|

jim
Genomic Map