Assay Details
| Gene Symbol: Arf79F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104197.3 | NP_001097667.1 | 3 - 4 | 511 | 131 | |
| NM_001104198.2 | NP_001097668.2 | 2 - 3 | 477 | 131 | ||
| NM_001275296.1 | NP_001262225.1 | 2 - 3 | 482 | 131 | ||
| NM_057607.6 | NP_476955.1 | 2 - 3 | 567 | 131 | ||
| NM_168970.4 | NP_730757.1 | 2 - 3 | 431 | 131 | ||
| NM_168971.4 | NP_730758.1 | 2 - 3 | 543 | 131 | ||
| NM_168972.5 | NP_730759.1 | 2 - 3 | 346 | 131 | ||
| NM_168973.3 | NP_730760.1 | 2 - 3 | 542 | 131 | ||
| GenBank mRNA | AY060375.1 | - | 2 - 3 | 360 | 131 | |
More Information
Set Membership: |
|

Arf79F
Genomic Map