Assay Details
| Gene Symbol: Naa60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144457.3 | NP_001137929.1 | 4 - 5 | 1178 | 71 | |
| NM_140096.5 | NP_648353.3 | 4 - 5 | 1178 | 71 | ||
| NM_206310.3 | NP_996032.1 | 3 - 4 | 1241 | 71 | ||
| GenBank mRNA | AY060437.1 | - | 3 - 4 | 1206 | 71 | |
| BT001491.1 | - | 4 - 5 | 1095 | 71 | ||
More Information
Set Membership: |
|

Naa60
Genomic Map