Assay Details
| Gene Symbol: CG6024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104117.2 | NP_001097587.1 | 4 - 5 | 1186 | 58 | |
| NM_001104119.2 | NP_001097589.2 | 4 - 5 | 1048 | 58 | ||
| NM_001382185.1 | NP_001368949.1 | 4 - 5 | 1186 | 58 | ||
| GenBank mRNA | BT021262.1 | - | 4 - 5 | 1186 | 58 | |
More Information
Set Membership: |
|

CG6024
Genomic Map