Assay Details
| Gene Symbol: CG6024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104117.2 | NP_001097587.1 | 5 - 6 | 1307 | 69 | |
| NM_001104119.2 | NP_001097589.2 | 5 - 6 | 1169 | 69 | ||
| NM_001382185.1 | NP_001368949.1 | 5 - 6 | 1307 | 69 | ||
| GenBank mRNA | BT021262.1 | - | 5 - 6 | 1307 | 69 | |
More Information
Set Membership: |
|

CG6024
Genomic Map