Assay Details
| Gene Symbol: app | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144464.2 | NP_001137936.1 | 9 - 10 | 2545 | 68 | |
| NM_001144465.2 | NP_001137937.1 | 8 - 9 | 2523 | 68 | ||
| NM_001144466.3 | NP_001137938.2 | 9 - 10 | 2545 | 68 | ||
| NM_001144467.3 | NP_001137939.2 | 8 - 9 | 2523 | 68 | ||
| NM_001259802.2 | NP_001246731.2 | 7 - 8 | 2142 | 68 | ||
| NM_001259803.2 | NP_001246732.1 | 8 - 9 | 2523 | 68 | ||
| NM_001259804.2 | NP_001246733.2 | 4 - 5 | 665 | 68 | ||
| NM_001300114.1 | NP_001287043.1 | 8 - 9 | 2491 | 68 | ||
| NM_140304.7 | NP_648561.2 | 8 - 9 | 2523 | 68 | ||
| NM_206343.5 | NP_996065.2 | 9 - 10 | 2545 | 68 | ||
| GenBank mRNA | BT030308.1 | - | 9 - 10 | 2493 | 68 | |
More Information
Set Membership: |
|

app
Genomic Map