Assay Details
| Gene Symbol: dlp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079347.3 | NP_524071.2 | 5 - 6 | 1607 | 65 | |
| NM_206353.2 | NP_996075.2 | 5 - 6 | 1607 | 65 | ||
| GenBank mRNA | AY052017.1 | - | 5 - 6 | 1588 | 65 | |
More Information
Set Membership: |
|

dlp
Genomic Map