Assay Details
| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140525.4 | NP_648782.2 | 4 - 5 | 732 | 66 | |
| NM_206375.3 | NP_996097.1 | 3 - 4 | 633 | 66 | ||
| GenBank mRNA | AY069537.1 | - | 3 - 4 | 574 | 66 | |
More Information
Set Membership: |
|

AIMP2
Genomic Map