Assay Details
| Gene Symbol: Taf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144486.3 | NP_001137958.1 | 9 - 10 | 3553 | 49 | |
| NM_001169982.2 | NP_001163453.1 | 9 - 10 | 3041 | 49 | ||
| NM_001274998.1 | NP_001261927.1 | 9 - 10 | 3175 | 49 | ||
| NM_001274999.1 | NP_001261928.1 | 9 - 10 | 3430 | 49 | ||
| NM_001300148.1 | NP_001287077.1 | 7 - 8 | 2645 | 49 | ||
| NM_079378.4 | NP_524102.1 | 9 - 10 | 3052 | 49 | ||
| NM_168651.3 | NP_730109.1 | 8 - 9 | 2993 | 49 | ||
| NM_176333.3 | NP_788511.1 | 8 - 9 | 2842 | 49 | ||
| NM_206379.3 | NP_996101.1 | 7 - 8 | 2585 | 49 | ||
| GenBank mRNA | BT025195.2 | - | 8 - 9 | 2790 | 49 | |
| S63550.1 | - | 8 - 9 | 2781 | 49 | ||
More Information
Set Membership: |
|

Taf4
Genomic Map