Assay Details
| Gene Symbol: CG43373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104152.4 | NP_001097622.2 | 19 - 20 | 3515 | 60 | |
| NM_168670.4 | NP_730173.3 | 19 - 20 | 3366 | 60 | ||
More Information
Set Membership: |
|

CG43373
Genomic Map