Assay Details
| Gene Symbol: CycT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275063.2 | NP_001261992.1 | 3 - 4 | 1931 | 77 | |
| NM_001275064.2 | NP_001261993.1 | 3 - 4 | 2162 | 77 | ||
| NM_079403.4 | NP_524127.2 | 3 - 4 | 1931 | 77 | ||
| NM_206395.3 | NP_996117.1 | 1 - 2 | 893 | 77 | ||
| GenBank mRNA | BT150266.1 | - | 3 - 4 | 1869 | 77 | |
More Information
Set Membership: |
|

CycT
Genomic Map