Assay Details
| Gene Symbol: Adgf-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079406.3 | NP_524130.1 | 3 - 4 | 1239 | 78 | |
| NM_206397.2 | NP_996119.1 | 3 - 4 | 1235 | 78 | ||
| GenBank mRNA | AF337554.1 | - | 3 - 4 | 1222 | 78 | |
| AY058655.1 | - | 3 - 4 | 1222 | 78 | ||
More Information
Set Membership: |
|

Adgf-A
Genomic Map