Assay Details
| Gene Symbol: arm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272220.1 | NP_001259149.1 | 3 - 4 | 1917 | 55 | |
| NM_057317.4 | NP_476665.2 | 3 - 4 | 1833 | 55 | ||
| NM_057318.4 | NP_476666.1 | 3 - 4 | 1826 | 55 | ||
| NM_134273.2 | NP_599100.1 | 3 - 4 | 1926 | 55 | ||
| NM_166912.2 | NP_726775.2 | 3 - 4 | 1926 | 55 | ||
| NM_206605.2 | NP_996328.1 | 3 - 4 | 1822 | 55 | ||
| GenBank mRNA | AF001213.1 | - | 2 - 3 | 1667 | 55 | |
| AY118525.1 | - | 3 - 4 | 1840 | 55 | ||
More Information
Set Membership: |
|

arm
Genomic Map