Assay Details
| Gene Symbol: arm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272220.1 | NP_001259149.1 | 6 - 7 | 2488 | 63 | |
| NM_057317.4 | NP_476665.2 | 6 - 7 | 2404 | 63 | ||
| NM_057318.4 | NP_476666.1 | 6 - 7 | 2397 | 63 | ||
| NM_134273.2 | NP_599100.1 | 6 - 7 | 2497 | 63 | ||
| NM_206605.2 | NP_996328.1 | 6 - 7 | 2393 | 63 | ||
| GenBank mRNA | AY118525.1 | - | 6 - 7 | 2411 | 63 | |
More Information
Set Membership: |
|

arm
Genomic Map