Assay Details
| Gene Symbol: eIF2Bepsilon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130605.5 | NP_569961.2 | 2 - 3 | 265 | 57 | |
| NM_206606.3 | NP_996329.1 | 2 - 3 | 265 | 57 | ||
| GenBank mRNA | AJ344150.1 | - | 2 - 3 | 237 | 57 | |
| AY095053.1 | - | 2 - 3 | 216 | 57 | ||
More Information
Set Membership: |
|

eIF2Bepsilon
Genomic Map