Assay Details
| Gene Symbol: sgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042794.4 | NP_001036259.2 | 8 - 9 | 2105 | 55 | |
| NM_001103400.2 | NP_001096870.1 | 8 - 9 | 1711 | 55 | ||
| NM_001169178.1 | NP_001162649.1 | 9 - 10 | 1730 | 55 | ||
| NM_001169179.1 | NP_001162650.1 | 8 - 9 | 1352 | 55 | ||
| NM_001272263.1 | NP_001259192.1 | 10 - 11 | 3233 | 55 | ||
| NM_001272264.1 | NP_001259193.1 | 9 - 10 | 3307 | 55 | ||
| NM_057366.5 | NP_476714.1 | 8 - 9 | 1737 | 55 | ||
| NM_057367.5 | NP_476715.1 | 8 - 9 | 1376 | 55 | ||
| NM_057368.5 | NP_476716.2 | 7 - 8 | 3334 | 55 | ||
| NM_134278.3 | NP_599105.1 | 8 - 9 | 1639 | 55 | ||
| NM_166947.3 | NP_726822.1 | 8 - 9 | 1584 | 55 | ||
| NM_166948.4 | NP_726823.1 | 8 - 9 | 1737 | 55 | ||
| NM_206611.2 | NP_996334.1 | 7 - 8 | 1293 | 55 | ||
| NM_206612.3 | NP_996335.1 | 7 - 8 | 1722 | 55 | ||
| NM_206613.3 | NP_996336.1 | 10 - 11 | 2026 | 55 | ||
| NM_206614.3 | NP_996337.1 | 9 - 10 | 1620 | 55 | ||
| NM_206615.2 | NP_996338.1 | 8 - 9 | 1550 | 55 | ||
| GenBank mRNA | AY122193.1 | - | 8 - 9 | 1712 | 55 | |
| BT050474.1 | - | 7 - 8 | 1672 | 55 | ||
| BT072831.1 | - | 8 - 9 | 1361 | 55 | ||
| BT133153.1 | - | 7 - 8 | 3328 | 55 | ||
| X70863.1 | - | 1696 | 55 | |||
More Information
Set Membership: |
|

sgg
Genomic Map