Assay Details
| Gene Symbol: sgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169178.1 | NP_001162649.1 | 10 - 11 | 2177 | 141 | |
| NM_001272263.1 | NP_001259192.1 | 11 - 12 | 3680 | 141 | ||
| NM_057367.5 | NP_476715.1 | 9 - 10 | 1823 | 141 | ||
| NM_057368.5 | NP_476716.2 | 8 - 9 | 3781 | 141 | ||
| NM_134278.3 | NP_599105.1 | 9 - 10 | 2086 | 141 | ||
| NM_166947.3 | NP_726822.1 | 9 - 10 | 2031 | 141 | ||
| NM_166948.4 | NP_726823.1 | 9 - 10 | 2184 | 141 | ||
| NM_206611.2 | NP_996334.1 | 8 - 9 | 1740 | 141 | ||
| NM_206612.3 | NP_996335.1 | 8 - 9 | 2169 | 141 | ||
| NM_206614.3 | NP_996337.1 | 10 - 11 | 2067 | 141 | ||
| NM_206615.2 | NP_996338.1 | 9 - 10 | 1997 | 141 | ||
| GenBank mRNA | AY122193.1 | - | 9 - 10 | 2159 | 141 | |
| BT050474.1 | - | 8 - 9 | 2119 | 141 | ||
| BT133153.1 | - | 8 - 9 | 3775 | 141 | ||
More Information
Set Membership: |
|

sgg
Genomic Map