Assay Details
| Gene Symbol: CG42541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038740.3 | NP_001033829.2 | 10 - 11 | 4235 | 65 | |
| NM_001272295.1 | NP_001259224.1 | 10 - 11 | 4235 | 65 | ||
| NM_001272296.1 | NP_001259225.1 | 11 - 12 | 4181 | 65 | ||
| NM_206622.4 | NP_996345.3 | 10 - 11 | 4238 | 65 | ||
More Information
Set Membership: |
|

CG42541
Genomic Map