Assay Details
| Gene Symbol: Vap33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130731.3 | NP_570087.1 | 4 - 5 | 898 | 80 | |
| NM_206623.2 | NP_996346.2 | 4 - 5 | 898 | 80 | ||
| NM_206624.2 | NP_996347.1 | 4 - 5 | 608 | 80 | ||
| NM_206625.2 | NP_996348.1 | 4 - 5 | 898 | 80 | ||
| GenBank mRNA | AY060395.1 | - | 4 - 5 | 869 | 80 | |
More Information
Set Membership: |
|

Vap33
Genomic Map