Assay Details
| Gene Symbol: CG34417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103423.3 | NP_001096893.1 | 11 - 12 | 10186 | 97 | |
| NM_001103425.3 | NP_001096895.1 | 11 - 12 | 10186 | 97 | ||
| NM_001169208.3 | NP_001162679.1 | 13 - 14 | 10660 | 97 | ||
| NM_001169209.3 | NP_001162680.1 | 12 - 13 | 16262 | 97 | ||
| NM_001201619.3 | NP_001188548.1 | 10 - 11 | 9915 | 97 | ||
| NM_001272347.1 | NP_001259276.1 | 10 - 11 | 5406 | 97 | ||
| NM_001272348.1 | NP_001259277.1 | 9 - 10 | 5055 | 97 | ||
| NM_001272349.1 | NP_001259278.1 | 8 - 9 | 2919 | 97 | ||
| NM_001272350.1 | NP_001259279.1 | 13 - 14 | 10634 | 97 | ||
| NM_001272351.1 | NP_001259280.1 | 12 - 13 | 15421 | 97 | ||
| NM_001272352.1 | NP_001259281.1 | 12 - 13 | 10531 | 97 | ||
| NM_001272353.1 | NP_001259282.1 | 12 - 13 | 15583 | 97 | ||
| NM_001272354.1 | NP_001259283.1 | 12 - 13 | 16265 | 97 | ||
| NM_001272355.1 | NP_001259284.1 | 11 - 12 | 10183 | 97 | ||
| NM_001298018.1 | NP_001284947.1 | 11 - 12 | 15991 | 97 | ||
| NM_001298019.1 | NP_001284948.1 | 9 - 10 | 9564 | 97 | ||
| NM_206636.2 | NP_996359.2 | 4 - 5 | 676 | 97 | ||
More Information
Set Membership: |
|

CG34417
Genomic Map