Assay Details
| Gene Symbol: Tom40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272384.1 | NP_001259313.1 | 3 - 4 | 898 | 54 | |
| NM_143746.5 | NP_652003.2 | 2 - 3 | 585 | 54 | ||
| NM_206640.2 | NP_996363.1 | 3 - 4 | 770 | 54 | ||
| GenBank mRNA | AY118926.1 | - | 2 - 3 | 562 | 54 | |
More Information
Set Membership: |
|

Tom40
Genomic Map