Assay Details
| Gene Symbol: fs(1)h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169228.2 | NP_001162699.1 | 4 - 5 | 2304 | 72 | |
| NM_078523.3 | NP_511078.2 | 2 - 3 | 1940 | 72 | ||
| NM_167144.4 | NP_727228.1 | 4 - 5 | 2405 | 72 | ||
| NM_206645.4 | NP_996368.1 | 3 - 4 | 2032 | 72 | ||
| NM_206646.4 | NP_996369.1 | 4 - 5 | 2330 | 72 | ||
| NM_206647.3 | NP_996370.1 | 4 - 5 | 2293 | 72 | ||
| GenBank mRNA | BT015270.1 | - | 4 - 5 | 2247 | 72 | |
| M23222.1 | - | 3 - 4 | 1965 | 72 | ||
More Information
Set Membership: |
|

fs(1)h
Genomic Map