Assay Details
| Gene Symbol: sdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038746.3 | NP_001033835.2 | 13 - 14 | 6771 | 69 | |
| NM_001038747.3 | NP_001033836.1 | 8 - 9 | 2791 | 69 | ||
| NM_001042798.2 | NP_001036263.1 | 6 - 7 | 2326 | 69 | ||
| NM_001258645.1 | NP_001245574.1 | 13 - 14 | 4658 | 69 | ||
| NM_001258646.1 | NP_001245575.1 | 4 - 5 | 2099 | 69 | ||
| NM_001258647.1 | NP_001245576.1 | 14 - 15 | 5997 | 69 | ||
| NM_001272397.1 | NP_001259326.1 | 13 - 14 | 6414 | 69 | ||
| NM_001272398.1 | NP_001259327.1 | 8 - 9 | 3959 | 69 | ||
| NM_001298081.1 | NP_001285010.1 | 12 - 13 | 5529 | 69 | ||
| NM_001298082.1 | NP_001285011.1 | 11 - 12 | 4230 | 69 | ||
| NM_132235.5 | NP_572463.1 | 9 - 10 | 4090 | 69 | ||
| NM_132236.6 | NP_572464.4 | 8 - 9 | 3523 | 69 | ||
| NM_206652.2 | NP_996375.1 | 4 - 5 | 2147 | 69 | ||
| NM_206653.3 | NP_996376.1 | 8 - 9 | 3752 | 69 | ||
| GenBank mRNA | BT010281.1 | - | 8 - 9 | 2789 | 69 | |
| BT099747.1 | - | 4 - 5 | 2147 | 69 | ||
More Information
Set Membership: |
|

sdt
Genomic Map