Assay Details
| Gene Symbol: Nrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169233.2 | NP_001162704.1 | 2 - 3 | 333 | 78 | |
| NM_001169234.3 | NP_001162705.1 | 2 - 3 | 364 | 78 | ||
| NM_001169235.1 | NP_001162706.1 | 2 - 3 | 338 | 78 | ||
| NM_001258652.1 | NP_001245581.1 | 2 - 3 | 338 | 78 | ||
| NM_001272407.2 | NP_001259336.1 | 2 - 3 | 1378 | 78 | ||
| NM_001272408.1 | NP_001259337.1 | 2 - 3 | 364 | 78 | ||
| NM_078535.3 | NP_511090.1 | 2 - 3 | 521 | 78 | ||
| NM_167160.2 | NP_727274.1 | 2 - 3 | 521 | 78 | ||
| NM_206657.4 | NP_996380.1 | 2 - 3 | 364 | 78 | ||
| GenBank mRNA | AY058284.1 | - | 2 - 3 | 521 | 78 | |
| M28231.2 | - | 1 - 2 | 93 | 78 | ||
More Information
Set Membership: |
|

Nrg
Genomic Map