Assay Details
| Gene Symbol: Nrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169233.2 | NP_001162704.1 | 7 - 8 | 3932 | 125 | |
| NM_001169235.1 | NP_001162706.1 | 7 - 8 | 3937 | 125 | ||
| NM_001272407.2 | NP_001259336.1 | 7 - 8 | 4977 | 125 | ||
| NM_078535.3 | NP_511090.1 | 7 - 8 | 4120 | 125 | ||
| NM_206657.4 | NP_996380.1 | 7 - 8 | 3963 | 125 | ||
| GenBank mRNA | AY058284.1 | - | 7 - 8 | 4120 | 125 | |
| M28231.2 | - | 6 - 7 | 3692 | 125 | ||
More Information
Set Membership: |
|

Nrg
Genomic Map