Assay Details
| Gene Symbol: Trf4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103443.2 | NP_001096913.1 | 3 - 4 | 1302 | 80 | |
| NM_001103444.2 | NP_001096914.1 | 3 - 4 | 1289 | 80 | ||
| NM_001272411.1 | NP_001259340.1 | 2 - 3 | 1594 | 80 | ||
| NM_132262.3 | NP_572490.2 | 2 - 3 | 1559 | 80 | ||
| NM_206658.4 | NP_996381.1 | 2 - 3 | 1721 | 80 | ||
| NM_206659.3 | NP_996382.2 | 2 - 3 | 1526 | 80 | ||
| NM_206660.2 | NP_996383.2 | 2 - 3 | 1296 | 80 | ||
| GenBank mRNA | BT010282.1 | - | 2 - 3 | 1408 | 80 | |
More Information
Set Membership: |
|

Trf4-1
Genomic Map