Assay Details
| Gene Symbol: Moe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258654.1 | NP_001245583.1 | 5 - 6 | 1365 | 92 | |
| NM_001258655.1 | NP_001245584.1 | 4 - 5 | 1431 | 92 | ||
| NM_001272432.1 | NP_001259361.1 | 6 - 7 | 1296 | 92 | ||
| NM_080343.4 | NP_525082.2 | 4 - 5 | 1304 | 92 | ||
| NM_167168.4 | NP_727290.1 | 5 - 6 | 1365 | 92 | ||
| NM_167169.2 | NP_727291.1 | 5 - 6 | 1256 | 92 | ||
| NM_167170.2 | NP_727292.1 | 1 - 2 | 930 | 92 | ||
| NM_206664.3 | NP_996387.1 | 4 - 5 | 1304 | 92 | ||
| NM_206665.3 | NP_996388.1 | 5 - 6 | 1194 | 92 | ||
| NM_206666.3 | NP_996389.1 | 4 - 5 | 1148 | 92 | ||
| NM_206667.3 | NP_996390.1 | 4 - 5 | 1207 | 92 | ||
| NM_206668.3 | NP_996391.1 | 4 - 5 | 1220 | 92 | ||
| NM_206669.2 | NP_996392.1 | 4 - 5 | 1195 | 92 | ||
| GenBank mRNA | AY069855.2 | - | 4 - 5 | 1288 | 92 | |
| BT099729.1 | - | 5 - 6 | 1365 | 92 | ||
More Information
Set Membership: |
|

Moe
Genomic Map