Assay Details
| Gene Symbol: AP-1gamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103448.3 | NP_001096918.1 | 8 - 9 | 1588 | 102 | |
| NM_001258660.1 | NP_001245589.1 | 7 - 8 | 1330 | 102 | ||
| NM_001258661.2 | NP_001245590.1 | 9 - 10 | 1606 | 102 | ||
| NM_001272442.1 | NP_001259371.1 | 8 - 9 | 1499 | 102 | ||
| NM_132299.6 | NP_572527.2 | 8 - 9 | 1357 | 102 | ||
| NM_167178.5 | NP_727311.2 | 7 - 8 | 1262 | 102 | ||
| NM_176717.4 | NP_788890.2 | 9 - 10 | 1375 | 102 | ||
| NM_176718.4 | NP_788891.2 | 8 - 9 | 1280 | 102 | ||
| NM_206671.4 | NP_996394.2 | 8 - 9 | 1295 | 102 | ||
| GenBank mRNA | BT100000.1 | - | 9 - 10 | 1601 | 102 | |
| BT100006.1 | - | 7 - 8 | 1331 | 102 | ||
| BT100128.1 | - | 8 - 9 | 1343 | 102 | ||
| BT133052.1 | - | 7 - 8 | 1260 | 102 | ||
More Information
Set Membership: |
|

AP-1gamma
Genomic Map