Assay Details
| Gene Symbol: alpha-Man-Ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167223.3 | NP_727407.1 | 2 - 3 | 1073 | 71 | |
| NM_206672.3 | NP_996395.1 | 2 - 3 | 1115 | 71 | ||
| NM_206673.3 | NP_996396.1 | 2 - 3 | 915 | 71 | ||
| NM_206674.3 | NP_996397.1 | 2 - 3 | 960 | 71 | ||
| NM_206675.3 | NP_996398.1 | 2 - 3 | 1068 | 71 | ||
| NM_206676.3 | NP_996399.1 | 2 - 3 | 892 | 71 | ||
| GenBank mRNA | BT011036.1 | - | 2 - 3 | 1072 | 71 | |
More Information
Set Membership: |
|

alpha-Man-Ia
Genomic Map