Assay Details
| Gene Symbol: alpha-Man-Ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272473.2 | NP_001259402.1 | 4 - 5 | 1674 | 70 | |
| NM_078550.4 | NP_511105.2 | 3 - 4 | 1499 | 70 | ||
| NM_167223.3 | NP_727407.1 | 4 - 5 | 1921 | 70 | ||
| NM_167224.3 | NP_727408.2 | 3 - 4 | 2057 | 70 | ||
| NM_206672.3 | NP_996395.1 | 4 - 5 | 1963 | 70 | ||
| NM_206673.3 | NP_996396.1 | 4 - 5 | 1763 | 70 | ||
| NM_206674.3 | NP_996397.1 | 4 - 5 | 1808 | 70 | ||
| NM_206675.3 | NP_996398.1 | 4 - 5 | 1916 | 70 | ||
| NM_206676.3 | NP_996399.1 | 4 - 5 | 1740 | 70 | ||
| GenBank mRNA | BT011036.1 | - | 4 - 5 | 1920 | 70 | |
| BT024451.1 | - | 3 - 4 | 2057 | 70 | ||
More Information
Set Membership: |
|

alpha-Man-Ia
Genomic Map