Assay Details
| Gene Symbol: alpha-Man-Ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272473.2 | NP_001259402.1 | 6 - 7 | 2068 | 67 | |
| NM_078550.4 | NP_511105.2 | 5 - 6 | 1893 | 67 | ||
| NM_167223.3 | NP_727407.1 | 6 - 7 | 2315 | 67 | ||
| NM_167224.3 | NP_727408.2 | 5 - 6 | 2451 | 67 | ||
| NM_206672.3 | NP_996395.1 | 6 - 7 | 2357 | 67 | ||
| NM_206673.3 | NP_996396.1 | 6 - 7 | 2157 | 67 | ||
| NM_206674.3 | NP_996397.1 | 6 - 7 | 2202 | 67 | ||
| NM_206675.3 | NP_996398.1 | 6 - 7 | 2310 | 67 | ||
| NM_206676.3 | NP_996399.1 | 6 - 7 | 2134 | 67 | ||
| GenBank mRNA | BT011036.1 | - | 6 - 7 | 2314 | 67 | |
| BT024451.1 | - | 5 - 6 | 2451 | 67 | ||
More Information
Set Membership: |
|

alpha-Man-Ia
Genomic Map