Assay Details
| Gene Symbol: PPP4R2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272481.2 | NP_001259410.1 | 3 - 4 | 617 | 68 | |
| NM_080344.3 | NP_525083.2 | 2 - 3 | 541 | 68 | ||
| NM_167232.2 | NP_727427.1 | 3 - 4 | 570 | 68 | ||
| NM_206677.2 | NP_996400.1 | 3 - 4 | 553 | 68 | ||
| GenBank mRNA | AY051757.1 | - | 3 - 4 | 566 | 68 | |
More Information
Set Membership: |
|

PPP4R2r
Genomic Map