Assay Details
| Gene Symbol: Gs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298192.1 | NP_001285121.1 | 3 - 4 | 768 | 66 | |
| NM_001298193.1 | NP_001285122.1 | 3 - 4 | 679 | 66 | ||
| NM_001298194.1 | NP_001285123.1 | 2 - 3 | 840 | 66 | ||
| NM_078568.3 | NP_511123.2 | 3 - 4 | 674 | 66 | ||
| NM_167284.2 | NP_727525.1 | 3 - 4 | 756 | 66 | ||
| NM_206685.2 | NP_996408.1 | 3 - 4 | 662 | 66 | ||
| GenBank mRNA | BT023772.1 | - | 3 - 4 | 733 | 66 | |
More Information
Set Membership: |
|

Gs2
Genomic Map