Assay Details
| Gene Symbol: CkIIbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014731.2 | NP_001014731.1 | 2 - 3 | 188 | 93 | |
| NM_001258699.1 | NP_001245628.1 | 2 - 3 | 107 | 93 | ||
| NM_001258700.2 | NP_001245629.1 | 2 - 3 | 476 | 93 | ||
| NM_001258701.2 | NP_001245630.1 | 2 - 3 | 210 | 93 | ||
| NM_001272533.1 | NP_001259462.1 | 2 - 3 | 476 | 93 | ||
| NM_078576.5 | NP_511131.3 | 2 - 3 | 142 | 93 | ||
| NM_080762.4 | NP_542940.1 | 2 - 3 | 229 | 93 | ||
| NM_167304.3 | NP_727561.1 | 2 - 3 | 476 | 93 | ||
| NM_167305.3 | NP_727562.1 | 2 - 3 | 111 | 93 | ||
| NM_206692.3 | NP_996415.1 | 2 - 3 | 229 | 93 | ||
| GenBank mRNA | AY113447.1 | - | 2 - 3 | 226 | 93 | |
| BT120031.1 | - | 2 - 3 | 445 | 93 | ||
| M16535.1 | - | 2 - 3 | 93 | 93 | ||
More Information
Set Membership: |
|

CkIIbeta
Genomic Map