Assay Details
| Gene Symbol: cac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014732.4 | NP_001014732.1 | 10 - 11 | 1695 | 61 | |
| NM_001014733.5 | NP_001014733.1 | 10 - 11 | 1695 | 61 | ||
| NM_001014734.4 | NP_001014734.1 | 10 - 11 | 1695 | 61 | ||
| NM_001014735.4 | NP_001014735.1 | 10 - 11 | 1695 | 61 | ||
| NM_001258707.2 | NP_001245636.1 | 10 - 11 | 1695 | 61 | ||
| NM_001258708.2 | NP_001245637.1 | 10 - 11 | 1695 | 61 | ||
| NM_001258709.2 | NP_001245638.1 | 9 - 10 | 1622 | 61 | ||
| NM_001258710.2 | NP_001245639.1 | 10 - 11 | 1695 | 61 | ||
| NM_001272548.1 | NP_001259477.1 | 9 - 10 | 1625 | 61 | ||
| NM_001370017.1 | NP_001356943.1 | 10 - 11 | 1695 | 61 | ||
| NM_001370018.1 | NP_001356944.1 | 10 - 11 | 1695 | 61 | ||
| NM_001370019.1 | NP_001356945.1 | 10 - 11 | 1695 | 61 | ||
| NM_078578.5 | NP_511133.2 | 10 - 11 | 1695 | 61 | ||
| NM_206693.5 | NP_996416.1 | 10 - 11 | 1695 | 61 | ||
| NM_206694.4 | NP_996417.1 | 10 - 11 | 1705 | 61 | ||
| NM_206695.4 | NP_996418.1 | 10 - 11 | 1695 | 61 | ||
| NM_206696.6 | NP_996419.1 | 10 - 11 | 1695 | 61 | ||
| NM_206697.4 | NP_996420.1 | 10 - 11 | 1695 | 61 | ||
More Information
Set Membership: |
|

cac
Genomic Map