Assay Details
| Gene Symbol: yl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078596.3 | NP_511151.2 | 1 - 2 | 535 | 71 | |
| NM_206710.2 | NP_996433.1 | 1 - 2 | 535 | 71 | ||
| GenBank mRNA | BT150470.1 | - | 1 - 2 | 530 | 71 | |
More Information
Set Membership: |
|

yl
Genomic Map