Assay Details
| Gene Symbol: Graf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298367.1 | NP_001285296.1 | 12 - 13 | 3450 | 70 | |
| NM_132842.3 | NP_573070.2 | 12 - 13 | 3423 | 70 | ||
| NM_206718.2 | NP_996441.2 | 13 - 14 | 3735 | 70 | ||
| NM_206719.2 | NP_996442.1 | 12 - 13 | 3650 | 70 | ||
| NM_206720.2 | NP_996443.1 | 13 - 14 | 3708 | 70 | ||
| NM_206721.2 | NP_996444.2 | 12 - 13 | 3677 | 70 | ||
| NM_206722.2 | NP_996445.1 | 12 - 13 | 3333 | 70 | ||
More Information
Set Membership: |
|

Graf
Genomic Map