Assay Details
| Gene Symbol: Graf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132842.3 | NP_573070.2 | 6 - 7 | 1268 | 129 | |
| NM_206719.2 | NP_996442.1 | 6 - 7 | 1495 | 129 | ||
| NM_206720.2 | NP_996443.1 | 7 - 8 | 1553 | 129 | ||
| NM_206722.2 | NP_996445.1 | 6 - 7 | 1178 | 129 | ||
More Information
Set Membership: |
|

Graf
Genomic Map