Assay Details
| Gene Symbol: Graf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298367.1 | NP_001285296.1 | 9 - 10 | 1960 | 120 | |
| NM_132842.3 | NP_573070.2 | 9 - 10 | 1933 | 120 | ||
| NM_206718.2 | NP_996441.2 | 10 - 11 | 2245 | 120 | ||
| NM_206719.2 | NP_996442.1 | 9 - 10 | 2160 | 120 | ||
| NM_206720.2 | NP_996443.1 | 10 - 11 | 2218 | 120 | ||
| NM_206721.2 | NP_996444.2 | 9 - 10 | 2187 | 120 | ||
| NM_206722.2 | NP_996445.1 | 9 - 10 | 1843 | 120 | ||
More Information
Set Membership: |
|

Graf
Genomic Map