Assay Details
| Gene Symbol: Stim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258774.2 | NP_001245703.1 | 4 - 5 | 1002 | 110 | |
| NM_078633.4 | NP_523357.2 | 4 - 5 | 938 | 110 | ||
| NM_206747.3 | NP_996470.2 | 4 - 5 | 938 | 110 | ||
| GenBank mRNA | AF328906.1 | - | 3 - 4 | 894 | 110 | |
| AY069686.1 | - | 4 - 5 | 922 | 110 | ||
| BT133089.1 | - | 4 - 5 | 916 | 110 | ||
More Information
Set Membership: |
|

Stim
Genomic Map