Assay Details
| Gene Symbol: Stim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258774.2 | NP_001245703.1 | 6 - 7 | 1288 | 67 | |
| NM_078633.4 | NP_523357.2 | 6 - 7 | 1224 | 67 | ||
| NM_206747.3 | NP_996470.2 | 6 - 7 | 1224 | 67 | ||
| GenBank mRNA | AF328906.1 | - | 5 - 6 | 1180 | 67 | |
| AY069686.1 | - | 6 - 7 | 1208 | 67 | ||
| BT133089.1 | - | 6 - 7 | 1202 | 67 | ||
More Information
Set Membership: |
|

Stim
Genomic Map