Assay Details
| Gene Symbol: Dsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144731.3 | NP_001138203.1 | 1 - 2 | 564 | 155 | |
| NM_001298393.1 | NP_001285322.1 | 3 - 4 | 605 | 155 | ||
| NM_080715.4 | NP_542446.2 | 3 - 4 | 1198 | 155 | ||
| NM_167502.3 | NP_727959.2 | 2 - 3 | 1262 | 155 | ||
| NM_167503.3 | NP_727960.1 | 3 - 4 | 677 | 155 | ||
| NM_167504.2 | NP_727961.1 | 3 - 4 | 602 | 155 | ||
| NM_206762.3 | NP_996485.1 | 3 - 4 | 746 | 155 | ||
| GenBank mRNA | BT126193.1 | - | 3 - 4 | 604 | 155 | |
More Information
Set Membership: |
|

Dsp1
Genomic Map